siRNA Details
Gene Symbol: Cpeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106276.1 | NP_001099746.1 | 2, 3 | 217 | ||
XM_008759509.1 | XP_008757731.1 | 2, 3 | 826 | |||
XM_008759510.1 | XP_008757732.1 | 2, 3 | 826 |
