siRNA Details
Gene Symbol: Tmem126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011557.1 | NP_001011557.1 | 3 | 289 | ||
XM_006229689.2 | XP_006229751.1 | 3 | 454 | |||
GenBank mRNA | BC089104.1 | - | 3 | 289 | 7307750 | |
FQ210414.1 | - | 3 | 310 | |||
FQ220582.1 | - | 3 | 448 | |||
FQ231598.1 | - | 3 | 339 |
