siRNA Details
Gene Symbol: Plcb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033350.1 | NP_203501.1 | 20 | 2429 | ||
XM_006230711.2 | XP_006230773.1 | 20 | 2561 | |||
GenBank mRNA | DQ120507.1 | - | 20 | 2429 | ||
DQ120508.1 | - | 20 | 2429 | |||
M99567.1 | - | 20 | 2363 |
