siRNA Details
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033889.1 | NP_001029061.1 | 14 | 1960 | ||
GenBank mRNA | BC101931.1 | - | 14 | 1960 | 7938773 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033889.1 | NP_001029061.1 | 14 | 1960 | ||
GenBank mRNA | BC101931.1 | - | 14 | 1960 | 7938773 |