siRNA Details
Gene Symbol: Xpo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011935.2 | NP_001011935.2 | 4 | 698 | ||
XM_008759870.1 | XP_008758092.1 | 4 | 803 | |||
GenBank mRNA | BC083625.1 | - | 4 | 698 |
