siRNA Details
Gene Symbol: Xpo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011935.2 | NP_001011935.2 | 24 | 3759 | ||
XM_008759870.1 | XP_008758092.1 | 24 | 3861 | |||
GenBank mRNA | CA504675.1 | - | 1 | 609 | ||
CV102769.1 | - | 5 | 708 |
