siRNA Details
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106295.1 | NP_001099765.1 | 10 | 1686 | ||
XM_006230224.1 | XP_006230286.1 | 10 | 1597 | |||
XM_006230225.1 | XP_006230287.1 | 9 | 1561 |
