siRNA Details
Gene Symbol: Cd2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106297.2 | NP_001099767.1 | 6 | 1012 | ||
XM_006230251.2 | XP_006230313.1 | 7 | 2774 | |||
XM_006230252.2 | XP_006230314.1 | 7 | 2782 | |||
XM_008759879.1 | XP_008758101.1 | 7 | 2788 | |||
GenBank mRNA | BC168174.1 | - | 6 | 1012 | 9032868 |
