siRNA Details
Gene Symbol: LOC103690122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759941.1 | XP_008758163.1 | 5 | 519 | ||
XM_008759942.1 | XP_008758164.1 | 5 | 519 | |||
XM_008759943.1 | XP_008758165.1 | 5 | 1292 | |||
XM_008759944.1 | XP_008758166.1 | 5 | 520 | |||
XM_008759945.1 | XP_008758167.1 | 5 | 519 | |||
XM_008759946.1 | XP_008758168.1 | 4 | 455 |
Gene Symbol: Ate1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106300.1 | NP_001099770.1 | 501 |
