siRNA Details
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277450.1 | NP_001264379.1 | 4 | 558 | ||
NM_019191.2 | NP_062064.1 | 4 | 586 | |||
XM_006254945.2 | XP_006255007.1 | 3 | 582 | |||
XM_006254946.2 | XP_006255008.1 | 4 | 299 | |||
GenBank mRNA | AB010147.1 | - | 3 | 360 | ||
AB017912.1 | - | 4 | 452 | |||
AF056001.1 | - | 4 | 486 | |||
BC127497.1 | - | 4 | 430 | 8364718 | ||
FQ230791.1 | - | 4 | 424 |
