siRNA Details
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277450.1 | NP_001264379.1 | 4, 5 | 731 | ||
NM_019191.2 | NP_062064.1 | 4, 5 | 759 | |||
XM_006254945.2 | XP_006255007.1 | 3, 4 | 755 | |||
XM_006254946.2 | XP_006255008.1 | 4, 5 | 472 | |||
GenBank mRNA | AB010147.1 | - | 3, 4 | 533 | ||
AB017912.1 | - | 4, 5 | 625 | |||
AF056001.1 | - | 4, 5 | 659 | |||
BC127497.1 | - | 4, 5 | 603 | 8364718 | ||
FQ230791.1 | - | 4, 5 | 597 |
