siRNA Details
Gene Symbol: Hrasls5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039007.1 | NP_001034096.1 | 3 | 846 | ||
XM_006230768.2 | XP_006230830.1 | 3 | 578 | |||
GenBank mRNA | AB255646.1 | - | 3 | 371 | ||
BC099084.1 | - | 3 | 846 | 7452977 |
