siRNA Details
Gene Symbol: Ccdc86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006974.2 | NP_001006975.1 | 1 | 566 | ||
GenBank mRNA | BC083770.1 | - | 1 | 584 | 7193350 | |
DQ501253.1 | - | 1 | 566 | |||
FQ213715.1 | - | 1 | 605 | |||
FQ219684.1 | - | 1 | 615 |
