siRNA Details
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135836.1 | NP_001129308.1 | 8 | 2067 | ||
XM_006229562.2 | XP_006229624.1 | 8 | 2626 | |||
XM_006229563.2 | XP_006229625.1 | 8 | 2477 | |||
XM_006229564.2 | XP_006229626.1 | 7 | 2563 | |||
XM_006229565.2 | XP_006229627.1 | 6 | 2281 |
