siRNA Details
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106353.1 | NP_001099823.1 | 5 | 358 | ||
XM_006231441.2 | XP_006231503.1 | 5 | 378 | |||
GenBank mRNA | BC161938.1 | - | 5 | 310 | 9022977 | |
FQ209728.1 | - | 5 | 379 | |||
FQ221472.1 | - | 4 | 243 |

More Information
Other Species Targeted

Gene Ontology Categories:
