siRNA Details
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106353.1 | NP_001099823.1 | 6 | 438 | ||
XM_006231441.2 | XP_006231503.1 | 6 | 458 | |||
GenBank mRNA | BC161938.1 | - | 6 | 390 | 9022977 | |
FQ209728.1 | - | 6 | 459 | |||
FQ221472.1 | - | 5 | 323 |
