siRNA Details
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011942.1 | NP_001011942.1 | 1 | 1696 | ||
XM_006231471.2 | XP_006231533.1 | 1 | 1789 | |||
XM_008760413.1 | XP_008758635.1 | 1 | 1790 | |||
GenBank mRNA | BC085930.1 | - | 1 | 1696 | 7110800 |
