siRNA Details
Gene Symbol: LOC100909771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749114.3 | XP_003749162.1 | 3 | 456 | ||
XM_006231376.2 | XP_006231438.1 | 3 | 456 | |||
XM_006231377.2 | XP_006231439.1 | 3 | 456 |
Gene Symbol: Ccnj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106369.1 | NP_001099839.1 | 421 | |||
XM_006231388.2 | XP_006231450.1 | 456 | ||||
XM_006231389.2 | XP_006231451.1 | 456 |
