siRNA Details
Gene Symbol: LOC100911660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231336.2 | XP_006231398.1 | 12 | 1731 | ||
XM_008760387.1 | XP_008758609.1 | 17 | 1999 | |||
XM_008760388.1 | XP_008758610.1 | 7 | 745 |
Gene Symbol: Hells | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106371.1 | NP_001099841.1 | 1970 | |||
XM_008760365.1 | XP_008758587.1 | 1731 | ||||
XM_008760366.1 | XP_008758588.1 | 7 | 745 |
