siRNA Details
Gene Symbol: March5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106372.2 | NP_001099842.1 | 2, 3 | 533 | ||
XM_008760367.1 | XP_008758589.1 | 2, 3 | 539 | |||
GenBank mRNA | BC158801.1 | - | 2, 3 | 533 | 7321043 |
