siRNA Details
Gene Symbol: Trim31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106376.1 | NP_001099846.1 | 1 | 292 | ||
XM_008772691.1 | XP_008770913.1 | 2 | 203 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106376.1 | NP_001099846.1 | 1 | 292 | ||
XM_008772691.1 | XP_008770913.1 | 2 | 203 |