siRNA Details
Gene Symbol: Msh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212536.2 | NP_997701.2 | 21 | 2022 | ||
XM_006256057.2 | XP_006256119.1 | 21 | 2852 | |||
XM_008772717.1 | XP_008770939.1 | 20 | 3093 | |||
XM_008772718.1 | XP_008770940.1 | 20 | 3156 | |||
GenBank mRNA | BC083904.1 | - | 21 | 2022 | 7133915 |
