siRNA Details
Gene Symbol: Slc44a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212541.1 | NP_997706.1 | 9 | 652 | ||
GenBank mRNA | BC079178.1 | - | 9 | 652 | 7130369 |

More Information
Other Species Targeted

Gene Ontology Categories:
