siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083586.2 | NP_001077055.2 | 2 | 425 | ||
NM_017197.3 | NP_058893.2 | 2 | 425 | |||
XM_008771861.1 | XP_008770083.1 | 2 | 304 | |||
XM_008771862.1 | XP_008770084.1 | 1 | 862 | |||
XM_008771863.1 | XP_008770085.1 | 1 | 861 | |||
XM_008771864.1 | XP_008770086.1 | 1 | 861 | |||
GenBank mRNA | AF090695.1 | - | 2 | 259 | ||
AJ010351.1 | - | 2 | 350 | |||
AJ010386.1 | - | 2 | 473 | |||
BC129096.1 | - | 2 | 251 | 8362847 |
