siRNA Details
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256148.1 | XP_006256210.1 | 3 | 329 | ||
XM_006256149.1 | XP_006256211.1 | 3 | 320 | |||
XM_006256151.2 | XP_006256213.1 | 2 | 241 | |||
XM_006256152.2 | XP_006256214.1 | 2 | 202 | |||
XM_228042.6 | XP_228042.4 | 2 | 538 |
