siRNA Details
Gene Symbol: Slc37a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011944.1 | NP_001011944.1 | 10 | 1170 | ||
XM_008772807.1 | XP_008771029.1 | 8 | 903 | |||
GenBank mRNA | BC081990.1 | - | 10 | 1170 | 7129340 |
