siRNA Details
Gene Symbol: Agpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106378.1 | NP_001099848.1 | 9 | 1494 | ||
XM_006256243.2 | XP_006256305.1 | 10 | 1289 | |||
XM_006256244.2 | XP_006256306.1 | 10 | 1317 | |||
GenBank mRNA | BC158873.1 | - | 8 | 1044 |
