siRNA Details
Gene Symbol: Aire | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106379.1 | NP_001099849.1 | 3 | 504 | ||
XM_006256247.2 | XP_006256309.1 | 3 | 571 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aire | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106379.1 | NP_001099849.1 | 3 | 504 | ||
XM_006256247.2 | XP_006256309.1 | 3 | 571 |