siRNA Details
Gene Symbol: Ranbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191604.1 | NP_001178533.1 | 7243 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ranbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191604.1 | NP_001178533.1 | 7243 |