siRNA Details
Gene Symbol: Hs3st5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106392.1 | NP_001099862.1 | 6 | 1567 | ||
XM_008772979.1 | XP_008771201.1 | 2 | 573 |

More Information
Other Species Targeted

Gene Ontology Categories:
