siRNA Details
Gene Symbol: Cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017202.1 | NP_058898.1 | 5 | 521 | ||
GenBank mRNA | BC084719.1 | - | 4 | 437 | 7317602 | |
FQ214265.1 | - | 5 | 530 | |||
FQ217878.1 | - | 5 | 489 | |||
FQ227128.1 | - | 5 | 519 | |||
FQ228805.1 | - | 5 | 471 | |||
X14209.1 | - | 5 | 503 | |||
X15029.1 | - | 4 | 448 |
