siRNA Details
Gene Symbol: Cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017202.1 | NP_058898.1 | 3 | 205 | ||
GenBank mRNA | BC084719.1 | - | 2 | 121 | 7317602 | |
FQ211185.1 | - | 3 | 158 | |||
FQ214265.1 | - | 3 | 214 | |||
FQ217878.1 | - | 3 | 173 | |||
FQ223981.1 | - | 3 | 167 | |||
FQ227128.1 | - | 3 | 203 | |||
FQ228805.1 | - | 3 | 155 | |||
X14209.1 | - | 3 | 187 | |||
X15029.1 | - | 2 | 132 |
