siRNA Details
Gene Symbol: Pald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034128.2 | NP_001029300.2 | 18 | 2290 | ||
XM_006256446.2 | XP_006256508.1 | 19 | 3065 | |||
XM_006256447.2 | XP_006256509.1 | 19 | 2501 | |||
GenBank mRNA | BC100093.1 | - | 2 | 89 |
