siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017448.1 | NP_001017448.1 | 8, 9 | 926 | ||
XM_006256562.2 | XP_006256624.1 | 8, 9 | 1207 | |||
XM_006256563.2 | XP_006256625.1 | 8, 9 | 1162 | |||
XM_006256564.2 | XP_006256626.1 | 8, 9 | 962 | |||
XM_006256565.1 | XP_006256627.1 | 9, 10 | 985 | |||
XM_006256566.2 | XP_006256628.1 | 8, 9 | 1619 | |||
XM_006256567.2 | XP_006256629.1 | 9, 10 | 954 | |||
XM_006256568.2 | XP_006256630.1 | 7, 8 | 837 | |||
XM_008772982.1 | XP_008771204.1 | 8, 9 | 1877 | |||
XM_008772983.1 | XP_008771205.1 | 8, 9 | 950 | |||
GenBank mRNA | BC079256.1 | - | 8, 9 | 926 | 7133287 |
