siRNA Details
Gene Symbol: Mcu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106398.1 | NP_001099868.1 | 7 | 897 |

More Information
Other Species Targeted

Gene Ontology Categories:

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106398.1 | NP_001099868.1 | 7 | 897 |