siRNA Details
Gene Symbol: Ttc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231719.2 | XP_006231781.1 | 5 | 367 | ||
XM_006231720.2 | XP_006231782.1 | 4 | 319 | |||
XM_006231721.2 | XP_006231783.1 | 4 | 349 | |||
XM_008760606.1 | XP_008758828.1 | 4 | 349 | |||
XM_008760607.1 | XP_008758829.1 | 5 | 387 | |||
XM_008760608.1 | XP_008758830.1 | 5 | 387 | |||
XM_008760609.1 | XP_008758831.1 | 4 | 349 | |||
XM_226606.8 | XP_226606.5 | 6 | 451 |
