siRNA Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100665.1 | NP_001094135.1 | 14, 15 | 1431 | ||
XM_006231830.2 | XP_006231892.1 | 15, 16 | 1607 | |||
GenBank mRNA | BC091392.1 | - | 12, 13 | 1222 |
