siRNA Details
Gene Symbol: Cd180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106405.1 | NP_001099875.1 | 3 | 727 | ||
XM_006231871.2 | XP_006231933.1 | 3 | 773 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106405.1 | NP_001099875.1 | 3 | 727 | ||
XM_006231871.2 | XP_006231933.1 | 3 | 773 |