siRNA Details
Gene Symbol: Cenpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106407.1 | NP_001099877.1 | 4 | 614 | ||
XM_006231888.1 | XP_006231950.1 | 4 | 614 | |||
XM_006231889.2 | XP_006231951.1 | 4 | 335 | |||
XM_006231890.2 | XP_006231952.1 | 4 | 246 | |||
XM_008760667.1 | XP_008758889.1 | 4 | 614 |
