siRNA Details
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106410.1 | NP_001099880.1 | 10 | 2135 | ||
XM_006231921.2 | XP_006231983.1 | 11 | 2501 | |||
XM_006231922.2 | XP_006231984.1 | 10 | 2441 | |||
XM_006231923.1 | XP_006231985.1 | 7 | 842 | |||
XM_008760725.1 | XP_008758947.1 | 6 | 782 | |||
GenBank mRNA | BC161907.1 | - | 9 | 1354 | 8367604 | |
FQ211661.1 | - | 11 | 2322 | |||
FQ213360.1 | - | 3 | 369 | |||
JQ013734.1 | - | 10 | 1221 |
