siRNA Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007633.2 | NP_001007634.1 | 4 | 433 | ||
NM_001162413.1 | NP_001155885.1 | 5 | 555 | |||
XM_006231985.2 | XP_006232047.1 | 5 | 501 | |||
XM_006231986.2 | XP_006232048.1 | 4 | 398 | |||
GenBank mRNA | BC079181.1 | - | 5 | 521 | 7130858 | |
CO554692.1 | - | 5 | 555 | |||
FM123325.1 | - | 4 | 387 | |||
FQ220090.1 | - | 5 | 450 | |||
FQ220210.1 | - | 4 | 346 | |||
FQ230755.1 | - | 4 | 356 | |||
FQ235152.1 | - | 5 | 483 |
