siRNA Details
Gene Symbol: Rai14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011947.1 | NP_001011947.1 | 14 | 2116 | ||
XM_006232060.2 | XP_006232122.1 | 15 | 2690 | |||
XM_006232061.2 | XP_006232123.1 | 15 | 2263 | |||
XM_006232062.2 | XP_006232124.1 | 15 | 3128 | |||
XM_006232063.2 | XP_006232125.1 | 14 | 2603 | |||
XM_006232064.2 | XP_006232126.1 | 13 | 2516 | |||
XM_008760765.1 | XP_008758987.1 | 13 | 3059 | |||
GenBank mRNA | BC085775.1 | - | 14 | 2116 | 7096978 |
