siRNA Details
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_215517.8 | XP_215517.5 | 20, 21 | 2188 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172606, AK129169, BC037454, AK132461, BC059190, and others.
Gene Ontology Categories:
