siRNA Details
Gene Symbol: Tnik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106422.1 | NP_001099892.1 | 21 | 2815 | ||
XM_008760893.1 | XP_008759115.1 | 20 | 3460 | |||
XM_008760894.1 | XP_008759116.1 | 20 | 3460 | |||
XM_008760895.1 | XP_008759117.1 | 19 | 3436 | |||
XM_008760896.1 | XP_008759118.1 | 19 | 3373 | |||
XM_008760897.1 | XP_008759119.1 | 18 | 3348 |
