siRNA Details
Gene Symbol: Nceh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127524.2 | NP_001120996.1 | 5 | 1179 | ||
GenBank mRNA | BC166533.1 | - | 5 | 1179 | 7190480 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nceh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127524.2 | NP_001120996.1 | 5 | 1179 | ||
GenBank mRNA | BC166533.1 | - | 5 | 1179 | 7190480 |