siRNA Details
Gene Symbol: Nceh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127524.2 | NP_001120996.1 | 1, 2 | 182 | ||
GenBank mRNA | BC166533.1 | - | 1, 2 | 182 | 7190480 |

More Information
Other Species Targeted

Gene Ontology Categories:
