siRNA Details
Gene Symbol: Padi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017227.1 | NP_058923.1 | 14, 15 | 1641 | ||
XM_006239145.2 | XP_006239207.1 | 14, 15 | 1665 | |||
GenBank mRNA | AB008803.1 | - | 14, 15 | 1641 | ||
AB010999.1 | - | 14, 15 | 1641 |
