siRNA Details
Gene Symbol: Ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106442.1 | NP_001099912.1 | 2, 3 | 241 | ||
GenBank mRNA | BC168754.1 | - | 2, 3 | 213 | 6886830 | |
FQ213377.1 | - | 2, 3 | 205 | |||
FQ221799.1 | - | 2, 3 | 179 | |||
FQ226826.1 | - | 2, 3 | 179 | |||
FQ229540.1 | - | 2, 3 | 213 | |||
FQ232929.1 | - | 2, 3 | 197 | |||
FQ235190.1 | - | 2, 3 | 197 |
