siRNA Details
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106449.2 | NP_001099919.1 | 4 | 461 | ||
NM_001270650.1 | NP_001257579.1 | 5 | 535 | |||
NM_001270651.1 | NP_001257580.1 | 5 | 508 | |||
NM_001270652.1 | NP_001257581.1 | 4 | 438 | |||
NM_001270653.1 | NP_001257582.1 | 4 | 465 | |||
XM_006232855.2 | XP_006232917.1 | 5 | 530 | |||
XM_008761282.1 | XP_008759504.1 | 4 | 496 | |||
GenBank mRNA | FN800735.1 | - | 4 | 419 | ||
FQ214903.1 | - | 4 | 398 | |||
FQ215988.1 | - | 5 | 464 | |||
FQ216125.1 | - | 4 | 387 | |||
FQ216236.1 | - | 2 | 132 | |||
FQ216249.1 | - | 5 | 458 | |||
FQ216534.1 | - | 4 | 364 | |||
FQ217203.1 | - | 4 | 385 | |||
FQ224853.1 | - | 4 | 386 |
