siRNA Details
Gene Symbol: Gabpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232936.2 | XP_006232998.1 | 8 | 1920 | ||
XM_227439.7 | XP_227439.6 | 8 | 1513 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232936.2 | XP_006232998.1 | 8 | 1920 | ||
XM_227439.7 | XP_227439.6 | 8 | 1513 |